Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD21 Antibody (544408), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347514 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347514 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347514 Supplier Novus Biologicals Supplier No. FAB4909MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD21 Monoclonal antibody specifically detects CD21 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CD21
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 544408
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human CD21, Ile21-Arg971, Accession # P20023
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Dendritic Cell Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.