Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD21 Antibody (Bu33), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10063574U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063574U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063574U Supplier Novus Biologicals Supplier No. NB10063574UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD21 Monoclonal specifically detects CD21 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD21
Applications Flow Cytometry
Classification Monoclonal
Clone Bu33
Conjugate DyLight 350
Dilution Flow Cytometry
Gene Accession No. P20023
Gene Alias C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Gene Symbols CR2
Host Species Mouse
Immunogen Made to Human CD21
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Dendritic Cell Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1380
Test Specificity NB100-63574 recognizes human CD21, a 145kD cell surface glycoprotein expressed by mature B lymphocytes, follicular dendritic cells, subsets of normal thymocytes and subsets of T cells. The CD21 molecule acts as a receptor for CD23, for complement components C3d, C3dg and iC3b and for Epstein Barr Virus. CD21 is part of the signalling complex that is linked to CD19, CD81 and phosphatidylinositol-3-kinase.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.