Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD21 Antibody (CR2/1952) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$658.00
Specifications
Antigen | CD21 |
---|---|
Clone | CR2/1952 |
Concentration | 1 mg/mL |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP26073301
![]() |
Novus Biologicals
NBP2607330.1MG |
0.1 mg |
Each for $658.00
|
|
|||||
NBP26073302
![]() |
Novus Biologicals
NBP2607330.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
CD21 Monoclonal specifically detects CD21 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
CD21 | |
1 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P20023 | |
1380 | |
Recombinant human CR2 (CD21) protein fragment (around aa 44-196) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. | |
140 kDa |
CR2/1952 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
Adaptive Immunity, Dendritic Cell Markers, Immunology | |
10mM PBS with No Preservative | |
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor | |
CR2 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 140kDa, which is identified as the complement receptor 2 (CR2)/ CD21. This protein is expressed strongly on mature B cells, follicular dendritic cells and weakly on immature thymocytes and T lymphocytes. In B-cell ontogeny, CD21 appears after the pre-B-stage, is maintained during peripheral B-cell development and is lost upon terminal differentiation into plasma cells. CD21 expression is also gradually lost after stimulation of B cells in vitro. CD21 functions as receptor for C3d, C3dg and iC3b Complement components, for EBV and for IFNalpha. CD21 binds to CD23 and associates with CD19, CD81 and Leu13 to form a large signal-transduction complex involved in B cell activation. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title