Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD217 (IL-17Ra) Monoclonal Antibody (424LTS), APC, eBioscience™
GREENER_CHOICE

Catalog No. p-7169261
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-153-31 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-153-31 Supplier Invitrogen™ Supplier No. 17791742
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The 424LTS monoclonal antibody reacts with and blocks IL-17A binding to human interleukin 17 receptor A (IL-17RA, IL-17R). IL-17RA is a type I membrane protein of approximately 96 kDa (866 amino acids) and is involved in inflammatory responses to IL-17A, IL-17F and IL-17AF heterodimer. It forms heterodimeric complexes with several other members of its receptor family, including IL-17RC for IL-17A and IL-17F signaling and IL-17RC for IL-17E (IL-25) signaling. IL-17RA is ubiquitously expressed, but in leukocytes its expression is elevated on granulocytes and monocytes. Applications Reported: This 424LTS antibody has been reported for use in flow cytometric analysis. Applications Tested: This 424LTS antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Interleukin 17A (IL-17A) is a proinflammatory cytokine secreted by activated T-lymphocytes. It is a potent inducer of the maturation of CD34-positive hematopoietic precursors into neutrophils. IL-17RA is a ubiquitous type I membrane glycoprotein that binds with low affinity to IL-17A and IL-17F. IL-17A and its receptor play a pathogenic role in many inflammatory and autoimmune diseases such as rheumatoid arthritis. Defects in IL-17RA are the cause of familial candidiasis type 5 (CANDF5) which is a rare disorder with altered immune responses and impaired clearance of fungal infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD217 (IL-17Ra)
Applications Flow Cytometry
Classification Monoclonal
Clone 424LTS
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Il17ra
Gene Accession No. Q96F46
Gene Alias AW538159; CANDF5; CD127; CD217; CDw217; hIL-17R; IL-17 receptor A; IL17R; Il17ra; IL-17RA; interleukin 17 receptor A; interleukin-17 receptor A; MGC10262; VDw217
Gene Symbols Il17ra
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23765
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.