Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (002), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB040288 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040288 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040288 Supplier Novus Biologicals Supplier No. NBP290617UV
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD229/SLAMF3/Lymphocyte Antigen 9
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 2
Conjugate DyLight 350
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD229/SLAMF3/Lymphocyte Antigen 9 (NP_032560.2; Met1-Arg453).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4063
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.