Learn More
CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (HLy9.1.25) - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CD229/SLAMF3/Lymphocyte Antigen 9 |
| Applications | Flow Cytometry, Immunoprecipitation, Functional Assay, CyTOF |
| Classification | Monoclonal |
| Clone | HLy9.1.25 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional, CyTOF-ready |
| Formulation | PBS with 0.09% Sodium Azide |
| Gene Alias | CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9 |
| Gene Symbols | LY9 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.