Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD229/SLAMF3/Lymphocyte Antigen 9 Antibody (MM0163-3L25), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP212151W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212151W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212151W Supplier Novus Biologicals Supplier No. NBP212151AF405

Mouse Monoclonal Antibody

CD229/SLAMF3/Lymphocyte Antigen 9 Monoclonal specifically detects CD229/SLAMF3/Lymphocyte Antigen 9 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD229/SLAMF3/Lymphocyte Antigen 9
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone MM0163-3L25
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry
Gene Accession No. Q9HBG7
Gene Alias CD229, CD229 antigen, cell-surface molecule Ly-9, hly9, lymphocyte antigen 9Cell surface molecule Ly-9, mLY9, SLAMF3, T-lymphocyte surface antigen Ly-9
Gene Symbols LY9
Host Species Mouse
Immunogen Human recombinant CD229/SLAMF3/Lymphocyte Antigen 9 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4063
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.