Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23/Fc epsilon RII Antibody (011), Alexa Fluor™ 405
SDP

Catalog No. NB076442 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB076442 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB076442 Supplier Novus Biologicals Supplier No. NBP290683AF405

Rabbit Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Mouse samples. It is validated for ELISA.

Specifications

Antigen CD23/Fc epsilon RII
Applications ELISA
Classification Monoclonal
Clone 11
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD23/Fc epsilon RII (P20693-1; Glu51-Pro331).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2208
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.