Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23/Fc epsilon RII Antibody (138628), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB175061 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175061 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175061 Supplier Novus Biologicals Supplier No. FAB123M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen CD23/Fc epsilon RII
Applications Blockade of Receptor-ligand Interaction, CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 138628
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD23/Fc epsilon RII, Met150-Ser321, Accession # P06734
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2208
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.