Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23/Fc epsilon RII Antibody (691632) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB375520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375520 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375520 Supplier Novus Biologicals Supplier No. FAB6900M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal antibody specifically detects CD23/Fc epsilon RII in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CD23/Fc epsilon RII
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 691632
Conjugate DyLight 594
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD23/Fc epsilon RII, Glu50-Pro331, Accession # P20693
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2208
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.