Learn More
CD23/Fc epsilon RII Antibody (EBVCS2), PerCP/Cy5.5, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP142855100TESTS
 View more versions of this product
Catalog No. NBP142855
Description
Description
CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD23/Fc epsilon RII
Monoclonal
PerCP-Cyanine5.5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
FCER2
The immunogen for this antibody was CD23.
100 Tests
Adaptive Immunity, Cancer, Immunology
2208
Store at 4C. Do not freeze.
Flow Cytometry
EBVCS2
Flow Cytometry 1:10-1:1000
BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Mouse
Affinity Purified
RUO
Primary
Human
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only