Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD23/Fc epsilon RII Antibody (EBVCS2), PerCP/Cy5.5, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | CD23/Fc epsilon RII |
|---|---|
| Clone | EBVCS2 |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP142855
![]() |
Novus Biologicals
NBP142855100TESTS |
100 Tests | N/A | N/A | N/A | ||||
Description
CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Flow Cytometry.Specifications
| CD23/Fc epsilon RII | |
| Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Mouse | |
| Adaptive Immunity, Cancer, Immunology | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
| 2208 | |
| The immunogen for this antibody was CD23. | |
| Primary | |
| Store at 4C. Do not freeze. |
| EBVCS2 | |
| Flow Cytometry | |
| PerCP-Cyanine5.5 | |
| RUO | |
| Human | |
| BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor | |
| FCER2 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title