Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD23/Fc epsilon RII Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169046 100 μg
NB169047 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB169046 Supplier Novus Biologicals Supplier No. NBP316985100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD23/Fc epsilon RII Polyclonal antibody specifically detects CD23/Fc epsilon RII in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen CD23/Fc epsilon RII
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias BLAST-2, CD23A, CD23CD23 antigen, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LKSQDLELSWNLNGLQADLSSFKSQELNERNEASDLLERLREEVTKLRMELQVSSGFVCNTCPEKWINFQRKCYYFGKGT
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2208
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.