Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Antibody (ML5), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB001049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001049 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001049 Supplier Novus Biologicals Supplier No. NB10077903G0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD24 Monoclonal specifically detects CD24 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen CD24
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ML5
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM Sodium Borate
Gene Alias CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Gene Symbols CD24
Host Species Mouse
Immunogen The immunogen was the full length version of the protein: CD24 (ML5)
Quantity 0.1 mL
Gene ID (Entrez) 100133941
Test Specificity CD24 (ML5)
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.