Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD24 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146390 0.1 mL
NBP146390SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146390 Supplier Novus Biologicals Supplier No. NBP146390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

CD24 Polyclonal specifically detects CD24 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CD24
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. P25063
Gene Alias CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen
Gene Symbols CD24
Host Species Rabbit
Immunogen A synthetic peptide from aa region 30-70 of human mature Signal transducer CD24 conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 100133941
Test Specificity Specific for CD24.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.