Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 5 publications
$244.50 - $499.50
Specifications
Antigen | CD24 |
---|---|
Dilution | Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP146390SS
![]() |
Novus Biologicals
NBP146390SS |
0.025 mL |
Each for $244.50
|
|
|||||
NBP146390
![]() |
Novus Biologicals
NBP146390 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
CD24 Polyclonal specifically detects CD24 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD24 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
P25063 | |
100133941 | |
A synthetic peptide from aa region 30-70 of human mature Signal transducer CD24 conjugated to blue carrier protein was used as the antigen. | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format. | |
Specific for CD24. |
Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
Cancer, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
Lyophilized from whole antisera with No Preservative | |
CD 24, CD24 antigen, CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24 molecule, CD24Asignal transducer CD24, FLJ22950, FLJ43543, MGC75043, Small cell lung carcinoma cluster 4 antigen | |
CD24 | |
IgG | |
Unpurified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title