Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2 R alpha Rat anti-Mouse, PE, Clone: AP-MAB0710, Novus Biologicals™
SDP

Catalog No. NB97867PE01 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB97867PE01 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB97867PE01 Supplier Novus Biologicals Supplier No. NB11097867PE0.1ML
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse, Rat samples. It is validated for Flow Cytometry, Product Image.

Specifications

Antigen CD25/IL-2 R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone AP-MAB0710
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Rat
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with recombinant mouse CD25 protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.