Learn More
CD25/IL-2Rα Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunofluorescence and flow cytometry experiments with Epredia™ CD25/IL-2Rα Ab-1, Mouse Monoclonal Antibody.
$197.00 - $646.00
Specifications
| Antigen | CD25/IL-2Ralpha Ab-1 |
|---|---|
| Clone | IL2R.1 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
|
MS203P0
|
Epredia
MS203P0 |
200μg/mL | 100 μL |
Each for $197.00
|
|
||||
|
MS203P1
|
Epredia
MS203P1 |
200μg/mL | 500 μL |
Each for $553.00
|
|
||||
|
MS203P1ABX
|
Epredia
MS203P1ABX |
1mg/mL | 100 μL |
Each for $646.00
|
|
||||
|
MS203P
|
Epredia
MS203P |
200μg/mL | 1 mL | N/A | N/A | N/A | |||
|
MS203PABX
|
Epredia
MS203PABX |
1mg/mL | 200 μL | N/A | N/A | N/A | |||
Description
IL-2R (T-cell growth factor receptor) consists of a chain of 55kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages.Host Species: Mouse
Clone: IL2R.1
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human interleukin-2 receptor
Molecular Weight: 55 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
| CD25/IL-2Ralpha Ab-1 | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| B-Cell | |
| Recombinant human interleukin-2 receptor | |
| Primary |
| IL2R.1 | |
| Monoclonal | |
| Concentrated | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.