Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD25/IL-2Rα Ab-4, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD25/IL-2Rα Ab-4, Mouse Monoclonal Antibody.

Specifications

Antigen CD25/IL-2Ralpha Ab-4
Clone 4C9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1088S1
View Documents
Epredia™
MS1088S1
Concentrated 500 μL N/A N/A N/A
MS1088S
View Documents
Epredia™
MS1088S
Concentrated 1 mL N/A N/A N/A
MS1088R1
View Documents
Epredia™
MS1088R1
Prediluted 1 mL N/A N/A N/A
MS1088R7
View Documents
Epredia™
MS1088R7
Prediluted 7 mL N/A N/A N/A
Description

Description

IL-2R (T-cell growth factor receptor) consists of a chain of 55 kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages. CD25 molecule alone appears to function as a low affinity receptor and associates wtih CD122 (IL-2R Β chain, p75) and CD132 (common γ chain) to form the high affinity IL-2 receptor complex

Host Species: Mouse

Clone: 4C9

Isotype: IgG2b

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein corresponding to the external domain of the Interleukin-2 Receptor molecule

Molecular Weight: 55 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD25/IL-2Ralpha Ab-4
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG2b
4C9
Monoclonal
Mouse
B-Cell
Recombinant protein corresponding to the external domain of the Interleukin-2 Receptor molecule
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.