Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2R alpha Antibody (280406), Biotin, Novus Biologicals™
SDP

Catalog No. NB365137 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365137 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365137 Supplier Novus Biologicals Supplier No. FAB2438B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD25/IL-2R alpha Monoclonal antibody specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CD25/IL-2R alpha
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 280406
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD25/IL-2 R alpha , Glu22-Lys236, Accession # P01590
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.