Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2R alpha Antibody (Daclizumab) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25266002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25266002 0.2 mg
NBP2526625 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25266002 Supplier Novus Biologicals Supplier No. NBP2526600.2MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD25/IL-2 R alpha Monoclonal specifically detects CD25/IL-2 R alpha in Cynomolgus Monkey, Human, Rhesus Macaque samples. It is validated for Block/Neutralize, CyTOF, Flow Cytometry.

Specifications

Antigen CD25/IL-2 R alpha
Applications Blocking Assay, CyTOF, Flow Cytometry
Classification Monoclonal
Clone Daclizumab
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Rabbit
Immunogen Cultured T-cells derived from a patient with Mycosis fungoides.
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3559
Test Specificity Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines.
Target Species Cynomolgus Monkey, Human, Rhesus Macaque
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.