Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25/IL-2R alpha Antibody (Daclizumab) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD25/IL-2 R alpha |
---|---|
Clone | Daclizumab |
Dilution | Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2526625
![]() |
Novus Biologicals
NBP2526600.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25266002
![]() |
Novus Biologicals
NBP2526600.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, CyTOF-ready, Block/Neutralize.Specifications
CD25/IL-2 R alpha | |
Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
IL2RA | |
IgG | |
Protein A or G purified | |
Binds to p55 chain of IL2R on human T cells, Tac antigen, which is expressed on T cells activated by mitogens, soluble antigens, and alloantigens but not on resting T cells, thymocytes, B cells, monocytes, activated B cells, leukemic T cell blasts from patients with acute lymphoblastic leukemia, and long-term T cell lines. |
Daclizumab | |
Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
3559 | |
Cultured T-cells derived from a patient with Mycosis fungoides. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title