Learn More
CD25/IL-2R alpha Antibody (OX-39), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CD25/IL-2R alpha |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OX-39 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR |
| Gene Symbols | IL2RA |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.