Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2R alpha Antibody (PC61.5.3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB001878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001878 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001878 Supplier Novus Biologicals Supplier No. NB600710G
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD25/IL-2R alpha
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone PC61.5.3
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Rat
Immunogen B6.1 CTL cell line.
Purification Method Protein G purified
Quantity 0.25 mL
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3559
Test Specificity This reacts with the low affinity alpha chain of the IL2 receptor present on activated T and B cells in mice. Clone PC61.5.3 is reported to inhibit IL2 binding and IL2 dependent proliferation.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.