Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25/IL-2R alpha Antibody (PC61.5.3), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB174477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174477 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174477 Supplier Novus Biologicals Supplier No. FAB9164Y
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CD25/IL-2R alpha
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone PC61.5.3
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 receptor subunit alpha, IL2R, IL-2R subunit alpha, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Host Species Rat
Immunogen B6.1 mouse cytotoxic T cell line, Accession # P01590
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3559
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.