Learn More
Invitrogen™ CD252 (OX40 Ligand) Monoclonal Antibody (RM134L), PerCP-eFluor™ 710, eBioscience™

Description
Description: The RM134L monoclonal antibody reacts with mouse CD252 also known as OX-40 Ligand, a member of the TNF superfamily. OX-40L is induced on mouse splenic B cells stimulated with a combination of anti-IgM and anti-CD40. Neither resting nor activated mouse T cells express OX-40L. The interaction of OX-40 (CD134) with OX-40L has been implicated in T cell-dependent humoral response. RM134L inhibits the costimulatory activity of mouse OX-40L transfected cells for anti-CD3-stimulated T cell proliferation. Applications Reported: This RM134L antibody has been reported for use in flow cytometric analysis. Applications Tested: This RM134L antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxida...
Specifications
Specifications
| Antigen | CD252 (OX40 Ligand) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RM134L |
| Concentration | 0.2 mg/mL |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Tnfsf4 |
| Gene Accession No. | P43488 |
| Gene Alias | Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.