Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD258 (LIGHT) Monoclonal Antibody (7-3 (7)), APC, eBioscience™

Mouse Monoclonal Antibody

$444.00

Specifications

Antigen CD258 (LIGHT)
Clone 7-3 (7)
Concentration 5 μL/Test
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Applications Flow Cytometry
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-151-34
Encompass_Preferred
View Documents
Invitrogen™
17258942
100 Tests
Each of 1 for $444.00
Only null left
Add to Cart
 
Description

Description

Description: This 7-3 (7) monoclonal antibody reacts with human CD258 (also known as LIGHT), a 29-kDa type II transmembrane protein that is a member of the TNF ligand superfamily related to lymphotoxin-β (LT-β). Expressed on activated T cells, particularly CD8+ T cells, LIGHT plays a role in cell survival, inflammation, and tumor eradication. Furthermore, upon binding its ligand, herpesvirus entry mediator (HVEM), LIGHT induces a costimulatory effect on T cells, enhancing T cell proliferation and cytokine production. LIGHT has also been shown to indirectly interact with the LT-β receptor. Applications Reported: This 7-3 (7) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 7-3 (7) antibody has been tested by flow cytometric analysis on PMA- and ionomycin-stimulated normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
Specifications

Specifications

CD258 (LIGHT)
5 μL/Test
Flow Cytometry
APC
Mouse
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
O43557
8740
Primary
Affinity Chromatography
7-3 (7)
4°C, store in dark, DO NOT FREEZE!
Monoclonal
Liquid
IgG1 κ
Human
TNFSF14
CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726
TNFSF14
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.