Learn More
Description
Host Species: Mouse
Clone: 137B4
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Immunogen: Recombinant human CD27 protein
Molecular Weight: 120 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD27 Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 137B4 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human CD27 protein |
| Purification Method | Tonsil |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.