Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ CD27 Ab-2, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD27 Ab-2, Mouse Monoclonal Antibody.
Specifications
| Antigen | CD27 Ab-2 |
|---|---|
| Clone | 137B4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
MS1468S0
|
Epredia™
MS1468S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
|
MS1468S1
|
Epredia™
MS1468S1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
|
MS1468R7
|
Epredia™
MS1468R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
CD27 (also known as S152 or T14) is expressed on the majority of peripheral T cells, medullary thymocytes, memory-type B cells, and natural killer cells. It is a transmembrane phosphoglycoprotein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. CD27 binds to its ligand CD70, a member of the TNF family, and induces T-cell costimulation and B-cell activation. It also interacts with TRAFs and is involved in activation of NFδB and SAPK/JNK and induces apoptosis.Host Species: Mouse
Clone: 137B4
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Immunogen: Recombinant human CD27 protein
Molecular Weight: 120 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
| CD27 Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG1 |
| 137B4 | |
| Monoclonal | |
| Mouse | |
| Apoptosis | |
| Recombinant human CD27 protein | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title