Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Ligand/TNFSF7/CD70 Antibody (301731), HRP, Novus Biologicals™
SDP

Catalog No. NB348789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348789 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348789 Supplier Novus Biologicals Supplier No. FAB2738H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD27 Ligand/TNFSF7/CD70
Applications Flow Cytometry
Classification Monoclonal
Clone 301731
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD27 Ligand/TNFSF7, Gln45-Pro193, Accession # AAA36175
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 970
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.