Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27 Ligand/TNFSF7/CD70 Antibody (TNFS7/1026), Novus Biologicals™
SDP

Catalog No. NBP24454600 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24454600 0.02 mg
NBP24454601 0.1 mg
NBP24454602 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24454600 Supplier Novus Biologicals Supplier No. NBP2445460.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunofluorescence

CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunofluorescence.

Specifications

Antigen CD27 Ligand/TNFSF7/CD70
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunofluorescence
Classification Monoclonal
Clone TNFS7/1026
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunoprecipitation 0.5 - 1 ug/500ug Protein Lysate, Immunohistochemistry-Frozen 0.5 - 1 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7
Gene Symbols CD70
Host Species Mouse
Immunogen Recombinant human CD70 protein
Molecular Weight of Antigen 30 kDa
Purification Method Protein A or G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 970
Test Specificity It recognizes a protein of 30kDa, identified as CD70. It is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T- and B-lymphocytes. It induces proliferation of co-stimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.