Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Antibody (137915), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB174116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174116 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174116 Supplier Novus Biologicals Supplier No. FAB5741Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Flow Cytometry, ELISA (Matched Antibody Pair)

Specifications

Antigen CD27/TNFRSF7
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 137915
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD27/TNFRSF7, Thr21-Arg182, Accession # P41272
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 939
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.