Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Antibody (618), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB333255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333255 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333255 Supplier Novus Biologicals Supplier No. NBP290425MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD27/TNFRSF7 Monoclonal antibody specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CD27/TNFRSF7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 618
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse CD27/TNFRSF7 (Accession#: NP_001028298.1, Met1-Arg182)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 939
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.