Learn More
Description
Specifications
Specifications
| Antigen | CD27/TNFRSF7 |
| Applications | Flow Cytometry, Immunoprecipitation, Functional Assay |
| Classification | Monoclonal |
| Clone | LG.7F9 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional |
| Formulation | Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide |
| Gene Alias | CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 |
| Gene Symbols | CD27 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
