Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Antibody (LG.7F9) - BSA Free, Novus Biologicals™
SDP

Armenian Hamster Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen CD27/TNFRSF7
Clone LG.7F9
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018549
SDP
View Documents
Novus Biologicals
NBP1434280.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP14342801
SDP
View Documents
Novus Biologicals
NBP1434280.1MG
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
NBP1434285
SDP
View Documents
Novus Biologicals
NBP1434280.5MG
0.5 mg N/A N/A N/A
Description

Description

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunoprecipitation.
Specifications

Specifications

CD27/TNFRSF7
0.5 mg/mL
Flow Cytometry, Functional Assay, Immunoprecipitation
Unconjugated
RUO
Human, Mouse, Rat
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
CD27
IgG
Affinity Purified
LG.7F9
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional
Monoclonal
Armenian Hamster
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
939
The immunogen for this antibody was CD27.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.