Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Antibody (O323), PE/Cy5, Novus Biologicals™
SDP

Catalog No. NBP143092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143092 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143092 Supplier Novus Biologicals Supplier No. NBP143092100TESTS

Mouse Monoclonal Antibody

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD27/TNFRSF7
Applications Flow Cytometry
Classification Monoclonal
Clone O323
Concentration 0.2 mg/mL
Conjugate PE-Cyanine5
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Gene Alias CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Gene Symbols CD27
Host Species Mouse
Immunogen The immunogen for this antibody was CD27.
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 939
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.