Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Antibody (RM27-3E5) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB001501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001501 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001501 Supplier Novus Biologicals Supplier No. NBP2751980.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Neutralization.

Specifications

Antigen CD27/TNFRSF7
Applications Flow Cytometry, ELISA, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone RM27-3E5
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, ELISA, Immunoprecipitation 1:10 - 1:500, Neutralization
Formulation PBS
Gene Alias CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Gene Symbols CD27
Host Species Rabbit
Immunogen This antibody was raised by immunising mouse with mouse CD27-human IgG1 Fc fusion protein. Isolated popliteal lymph node cells were then fused with P3U1 myeloma cells to produce stable hybridomas.
Quantity 0.025 mg
Gene ID (Entrez) 939
Test Specificity This antibody is specific for the extracellular domain of murine CD27, a TNF-receptor superfamily member.
Target Species Mouse
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.