Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD272 (BTLA) Monoclonal Antibody (MIH26), Functional Grade, eBioscience™
GREENER_CHOICE

Catalog No. 5014659
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-146-59 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-146-59 Supplier Invitrogen™ Supplier No. 16597982
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The BTLA monoclonal antibody reacts with human B and T lymphocyte attenuator (BTLA). BTLA is expressed by peripheral lymphocytes, splenic macrophages, developing B cells in the bone marrow and developing T cells in the thymus and mature, but not immature bone marrow-derived dendritic cells. BTLA is implicated as a negative regulator of the activation and/or function of various hemopoietic cell types. It is reported that BTLA binds to B7-H4, but further studies are needed to confirm this interaction. Applications Reported: This MIH26 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH26 antibody has been tested by flow cytometric analysis of normal human peripheral blood. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

BTLA or B and T-lymphocyte attenuator is a member of the co-inhibitory receptors of the CD28 superfamily. BTLA is a lymphocyte inhibitory receptor which inhibits lymphocytes during immunes responses. BTLA is constitutively expressed on most CD4+ and CD8+ T cells and its expression progressively decreases upon T cell activation. It remains expressed on Th1 cells, but not Th2 cells. BTLA is a unique co-receptor that interacts with the tumor necrosis factor receptor superfamily member herpesvirus entry mediator (HVEM). This interation is an important pathway regulating lymphocyte activation and/or homeostasis in the immune response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD272 (BTLA)
Applications Control, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone MIH26
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene BTLA
Gene Accession No. Q7Z6A9
Gene Alias A630002H24; B and T lymphocyte associated; B and T lymphocyte associated protein; B and T lymphocyte attenuator; B- and T-lymphocyte attenuator; B- and T-lymphocyte-associated protein; BTLA; BTLA_HUMAN; BTLA1; CD272; CD272 antigen; FLJ16065; MGC129743
Gene Symbols BTLA
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151888
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.