Learn More
CD276 Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA813302

Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
CD276 Monoclonal antibody specifically detects CD276 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
CD276 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5ZPR3, Q7TPB4, Q8VE98 | |
Mouse | |
Protein A/G | |
RUO | |
102657, 315716, 80381 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
OTI1A10 | |
Unconjugated | |
CD276 | |
4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
Full length human recombinant protein of human CD276 (NP_079516) produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.