Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD276 Monoclonal Antibody (OTI1A10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$516.29
Specifications
Antigen | CD276 |
---|---|
Clone | OTI1A10 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-224-4488
![]() ![]() |
OriGene
TA813302 |
100 μL |
Each of 1 for $516.29
|
|
|||||
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
CD276 Monoclonal antibody specifically detects CD276 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
CD276 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352 | |
Full length human recombinant protein of human CD276 (NP_079516) produced in HEK293T cell. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
CD276 |
OTI1A10 | |
Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q5ZPR3, Q7TPB4, Q8VE98 | |
102657, 315716, 80381 | |
IgG1 | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title