Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD277/BTN3A1 Antibody (BT3.1), FITC, Novus Biologicals™
SDP

Catalog No. NB355767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355767 Supplier Novus Biologicals Supplier No. NBP200227F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD277/BTN3A1 Monoclonal antibody specifically detects CD277/BTN3A1 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation

Specifications

Antigen CD277/BTN3A1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone BT3.1
Conjugate FITC
Dilution Flow Cytometry, Immunoprecipitation
Formulation PBS
Gene Alias butyrophilin, subfamily 3, member A1, dJ45P21.3 (butyrophilin, subfamily 3, member A1)
Host Species Mouse
Immunogen The immunogen for this antibody is CD277 (B7 family, Butyrophilin).
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11119
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.