Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 506, eBioscience™, Invitrogen™

Catalog No. p-7111198
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69998580 25 μg
69998582 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 69998580 Supplier Invitrogen™ Supplier No. 69998580
Only null left
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

700012 has successfully been used in western blot to detect phospho-ERK1/2 in TPA-treated HeLa, NRK and MDCK cell lysates. This antibody is predicted to react with bovine, chicken, chimpanzee, mouse, rat, Xenopus and zebrafish based on sequence homology. ABfinity™ recombit antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale, and purified with Protein A. ABfinity™ monoclonal antibodies resemble rabbit monoclonals isolated from serum or produced by hybridomas, but demonstrate greater specificity and sensitivity. Because ABfinity™ recombit antibodies are derived from cloned DNA sequences of the heavy and light antibody chains, they are not susceptible to cell-line drift or lot-to-lot variation, thus allowing for peak specificity and performance. Intact IgG appears on a non-reducing gel as ∽150 kDa band and upon reduction generating a ∽25 kDa light chain band and a ∽50 kDa heavy chain.

Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of 'danger signals' such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD279 (PD-1)
Applications Flow Cytometry
Classification Monoclonal
Clone J43
Concentration 0.2 mg/mL
Conjugate eFluor 506
Formulation PBS with 0.09% sodium azide
Gene Pdcd1
Gene Accession No. Q02242
Gene Alias CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2
Gene Symbols Pdcd1
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18566
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.