Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Antibody (37407), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB304003 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304003 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304003 Supplier Novus Biologicals Supplier No. FAB342Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD28 Monoclonal antibody specifically detects CD28 in Human samples. It is validated for Western Blot, Agonist Activity, Immunocytochemistry

Specifications

Antigen CD28
Applications Western Blot, Agonist Activity, Immunocytochemistry
Classification Monoclonal
Clone 37407
Conjugate DyLight 680
Dilution Western Blot, Agonist Activity, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CD28, Asn19-Pro152, Accession # P10747
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 940
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.