Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Antibody (CB28), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP234592G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234592G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234592G Supplier Novus Biologicals Supplier No. NBP234592G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD28
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CB28
Conjugate DyLight 488
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Gene Alias CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Gene Symbols CD28
Host Species Mouse
Immunogen Recombinant human CD28 protein (Uniprot: P10747)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 940
Test Specificity Recognizes a glycoprotein of 44-88kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.