Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Antibody (PV-1), Biotin, Novus Biologicals™
SDP

Catalog No. NBP127934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP127934 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP127934 Supplier Novus Biologicals Supplier No. NBP127934
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD28 Monoclonal specifically detects CD28 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, In vitro assay.

Specifications

Antigen CD28
Applications Flow Cytometry, Immunoprecipitation, In vitro Assay
Classification Monoclonal
Clone PV-1
Concentration 0.5 mg/mL
Conjugate Biotin
Dilution Flow Cytometry <= 3 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay
Gene Alias CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Gene Symbols CD28
Host Species Armenian Hamster
Immunogen C57BL/6N mouse T cell lymphoma EL4 cell line
Purification Method Protein A or G purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 940
Test Specificity Mouse CD28
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.