Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD28 Antibody (YTH913.12), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB064867549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064867549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064867549 Supplier Novus Biologicals Supplier No. NB10064867JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD28 Monoclonal specifically detects CD28 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen CD28
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone YTH913.12
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Gene Symbols CD28
Host Species Rat
Immunogen Human peripheral blood T-cells
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 940
Test Specificity NB100-64867 recognizes the human CD28 cell surface antigen, a 44kD glycoprotein expressed by a subset of T lymphocytes. The antibody enhances the MLR response. It has been reported that this clone also recognizes an epitope of CD28 expressed by NK cells, which is not recognized by clones of 9.3 or CD28.2.(2)
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.