Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD28, Mouse Anti-Human, RY610, Clone: CD28.2, BD Horizon™
Mouse Monoclonal Antibody
$186.00 - $370.00
Specifications
Antigen | CD28 |
---|---|
Clone | CD28.2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RY610 |
Description
CD28 Monoclonal antibody specifically detects CD28 in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow CytometrySpecifications
CD28 | |
Flow Cytometry | |
RY610 | |
Mouse | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
CD28 antigen; T44; Tp44; TP44 | |
Human CD28 Transfected Cell Line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
CD28.2 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
940, 705313, 100126740, 102146670 | |
IgG1 κ | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title