Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2F-10/SLAMF9 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB204665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB204665 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB204665 Supplier Novus Biologicals Supplier No. NBP176534AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD2F-10/SLAMF9 Polyclonal antibody specifically detects CD2F-10/SLAMF9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CD2F-10/SLAMF9
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human SLAMF9. The immunogen is located within the last 50 amino acids of SLAMF9. Amino Acid Squence: RMKKLMRNRMKLRKEAKPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 89886
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.