Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Antibody (17A2), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB368966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368966 Supplier Novus Biologicals Supplier No. FAB4841I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD3 Monoclonal specifically detects CD3 in Mouse samples. It is validated for Complement-dependent Cytotoxicity, CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, T Cell Stimulation.

Specifications

Antigen CD3
Applications Complement-dependent Cytotoxicity, CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone 17A2
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity, T Cell Stimulation, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD_antigen: CD3e, CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, IMD18, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Host Species Rat
Immunogen T cell hybridoma D1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 916
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.