Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Antibody (KT3) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064871 0.125 mg
NB017658 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064871 Supplier Novus Biologicals Supplier No. NB10064871
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD3 Monoclonal specifically detects CD3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen CD3
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone KT3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Gene Symbols CD3
Host Species Rat
Immunogen CBAT6 thymocytes
Molecular Weight of Antigen 21.4 kDa
Purification Method Protein G purified
Quantity 0.125 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 916
Test Specificity The antibody recognizes mouse CD3 antigen.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.